CO1H324A63BB CombiPhos Catalysts Inc

2-Carbamoylthiazole-4-boronic acid pinacol ester

  • Cas Number : 2223044-61-7
  • Molecular Weight : 254.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart