CO1H324A63DA CombiPhos Catalysts Inc

4-(4-Bromothiazol-5-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223050-78-8
  • Molecular Weight : 366.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart