CO1H324A63E2 CombiPhos Catalysts Inc

4-(3-Chlorophenyl)thiazole-5-boronic acid pinacol ester

  • Cas Number : 2223039-42-5
  • Molecular Weight : 321.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart