CO1H324A63EB CombiPhos Catalysts Inc

3-(tert-Butyl)-5-(3-furyl)phenylboronic acid

  • Cas Number : 2225169-55-9
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart