CO1H324A63EF CombiPhos Catalysts Inc

2-(5-cyclopentylfuran-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 2223053-58-3
  • Molecular Weight : 262.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart