CO1H324A63F2 CombiPhos Catalysts Inc

5-Methyl-N-Boc-imidazole-4-boronic acid pinacol ester

  • Cas Number : 1402174-59-7
  • Molecular Weight : 308.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart