CO1H324A63F5 CombiPhos Catalysts Inc

tert-butyl 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole-1-carboxylate

  • Cas Number : 1402174-60-0
  • Molecular Weight : 308.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart