CO1H324A63F6 CombiPhos Catalysts Inc

N-Boc-5-methylpyrazole-3-boronic acid pinacol ester

  • Cas Number : 1402174-61-1
  • Molecular Weight : 308.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart