CO1H324A6404 CombiPhos Catalysts Inc

Pyrrole-2-boronic acid pinacol ester

  • Cas Number : 476004-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart