CO1H324A640C CombiPhos Catalysts Inc

N-(2-Acetoxyethyl)-2-methylimidazole-4-boronic acid pinacol ester

  • Cas Number : 1402166-63-5
  • Molecular Weight : 294.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart