CO1H324A6415 CombiPhos Catalysts Inc

4,4,5,5-tetramethyl-2-(4-propylfuran-2-yl)-1,3,2-dioxaborolane

  • Cas Number : 2223030-87-1
  • Molecular Weight : 236.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart