CO1H324A641A CombiPhos Catalysts Inc

4-(2-Thienyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223031-54-5
  • Molecular Weight : 276.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart