CO1H324A6422 CombiPhos Catalysts Inc

5-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)FURAN-2-CARBONITRILE

  • Cas Number : 1111096-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart