CO1H324A642C CombiPhos Catalysts Inc

5-Aminopiperazine-2-boronic acid pinacol ester

  • Cas Number : 2223009-22-9
  • Molecular Weight : 227.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart