CO1H324A6435 CombiPhos Catalysts Inc

2-(n-Propyl)thiophene-4-boronic acid pinacol ester

  • Cas Number : 1595111-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart