CO1H324A6436 CombiPhos Catalysts Inc

2-(iso-Propyl)thiophene-4-boronic acid pinacol ester

  • Cas Number : 2223050-93-7
  • Molecular Weight : 252.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart