CO1H324A6437 CombiPhos Catalysts Inc

2-(sec-Butyl)thiophene-4-boronic acid pinacol ester

  • Cas Number : 2223006-06-0
  • Molecular Weight : 266.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart