CO1H324A643F CombiPhos Catalysts Inc

2-Methyl-5-ethylthiophene-3-boronic acid pinacol ester

  • Cas Number : 2223031-03-4
  • Molecular Weight : 252.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart