CO1H324A6445 CombiPhos Catalysts Inc

2-Methyl-5-(sec-butyl)thiophene-3-boronic acid pinacol ester

  • Cas Number : 2223006-79-7
  • Molecular Weight : 280.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart