CO1H324A6449 CombiPhos Catalysts Inc

2-Methyl-5-(tert-butyl)thiophene-3-boronic acid pinacol ester

  • Cas Number : 2223039-48-1
  • Molecular Weight : 280.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart