CO1H324A6465 CombiPhos Catalysts Inc

6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile

  • Cas Number : 1073353-83-9
  • Molecular Weight : 230.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile
Inchi Code 1S/C12H15BN2O2/c1-11(2)12(3,4)17-13(16-11)10-6-5-9(7-14)8-15-10/h5-6,8H,1-4H3
Inchi Key USYWPRFJTBASTH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart