CO1H324A6467 CombiPhos Catalysts Inc

5-(1H-Pyrazol-1-yl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223043-06-7
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart