CO1H324A646F CombiPhos Catalysts Inc

5-(2-Methylimidazol-1-yl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223042-27-9
  • Molecular Weight : 274.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart