CO1H324A6479 CombiPhos Catalysts Inc

5-(Piperidino)furan-2-boronic acid pinacol ester

  • Cas Number : 2223051-14-5
  • Molecular Weight : 277.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart