CO1H324A6488 CombiPhos Catalysts Inc

4-(3-Methyl-1H-pyrazol-1-yl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 2223043-28-3
  • Molecular Weight : 290.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart