CO1H324A648D CombiPhos Catalysts Inc

4-(Imidazol-1-yl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223050-07-3
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart