CO1H324A6497 CombiPhos Catalysts Inc

4-(Morpholino)furan-2-boronic acid pinacol ester

  • Cas Number : 2223042-47-3
  • Molecular Weight : 279.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart