CO1H324A649E CombiPhos Catalysts Inc

4-(4-Methylpiperidin-1-yl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 2223031-88-5
  • Molecular Weight : 307.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart