CO1H324A64A2 CombiPhos Catalysts Inc

4-(Diethylamino)thiophene-2-boronic acid pinacol ester

  • Cas Number : 2223032-38-8
  • Molecular Weight : 281.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart