CO1H324A64A6 CombiPhos Catalysts Inc

5-(N-Methylpiperazin-1-yl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 2223033-64-3
  • Molecular Weight : 308.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart