CO1H324A64AD CombiPhos Catalysts Inc

2,5-Dimethoxy-2,5-dihydrofuran-3-boronic acid pinacol ester

  • Cas Number : 2223003-40-3
  • Molecular Weight : 256.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart