CO1H324A64B6 CombiPhos Catalysts Inc

4-(Acetyl)furan-2-boronic acid pinacol ester

  • Cas Number : 1111096-31-1
  • Molecular Weight : 236.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart