CO1H324A64B8 CombiPhos Catalysts Inc

3-(Acetyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223029-74-9
  • Molecular Weight : 236.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart