CO1H324A64BC CombiPhos Catalysts Inc

3-Cyclobutylthiophene-2-boronic acid pinacol ester

  • Cas Number : 2223040-29-5
  • Molecular Weight : 264.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart