CO1H324A64BE CombiPhos Catalysts Inc

3-Cyclopentylfuran-2-boronic acid pinacol ester

  • Cas Number : 2223042-57-5
  • Molecular Weight : 262.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart