CO1H324A64BF CombiPhos Catalysts Inc

3-(Oxolan-3-yl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 2223009-31-0
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart