CO1H324A64C1 CombiPhos Catalysts Inc

3-Cyclohexylthiophene-2-boronic acid pinacol ester

  • Cas Number : 2223051-24-7
  • Molecular Weight : 292.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart