CO1H324A64D0 CombiPhos Catalysts Inc

1-Acetyl-2-(iso-propyl)imidazole-4-boronic acid pinacol ester

  • Cas Number : 2223029-85-2
  • Molecular Weight : 278.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart