CO1H324A64D1 CombiPhos Catalysts Inc

2-isopropyl-1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole

  • Cas Number : 2223040-44-4
  • Molecular Weight : 250.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart