CO1H324A64DD CombiPhos Catalysts Inc

tert-butyl 3-(2-isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazol-1-yl)piperidine-1-carboxylate

  • Cas Number : 2223003-68-5
  • Molecular Weight : 419.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart