CO1H324A64E2 CombiPhos Catalysts Inc

2-METHYL-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-1H-PYRROLE

  • Cas Number : 676501-88-5
  • Molecular Weight : 207.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart