CO1H324A64EC CombiPhos Catalysts Inc

2,5-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-pyrrole

  • Cas Number : 2223047-94-5
  • Molecular Weight : 221.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart