CO1H324A64ED CombiPhos Catalysts Inc

1-Methyl-5-acetylpyrrole-3-boronic acid pinacol ester

  • Cas Number : 2223043-85-2
  • Molecular Weight : 249.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart