CO1H324A64EE CombiPhos Catalysts Inc

5-Acetyl-1H-pyrrole-2-boronic acid pinacol ester

  • Cas Number : 2223040-19-3
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart