CO1H324A64F4 CombiPhos Catalysts Inc

3,5-Dimethyl-1H-pyrrole-2-boronic acid pinacol ester

  • Cas Number : 2223032-92-4
  • Molecular Weight : 221.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart