CO1H324A64FA CombiPhos Catalysts Inc

2-(N-Boc-Piperidin-3-yl)thiazole-5-boronic acid neopentylglycol ester

  • Cas Number : 2223002-81-9
  • Molecular Weight : 380.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart