CO1H324A64FC CombiPhos Catalysts Inc

2-(Oxan-4-yl)thiazole-5-boronic acid neopentylglycol ester

  • Cas Number : 2223032-01-5
  • Molecular Weight : 281.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart