CO1H324A6501 CombiPhos Catalysts Inc

5-Chloropyrazine-2-boronic acid neopentylglycol ester

  • Cas Number : 2223046-36-2
  • Molecular Weight : 226.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart