CO1H324A650D CombiPhos Catalysts Inc

1-isobutyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole

  • Cas Number : 2223040-61-5
  • Molecular Weight : 249.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart