CO1H324A6512 CombiPhos Catalysts Inc

1-cyclopropyl-2-ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole

  • Cas Number : 2223030-25-7
  • Molecular Weight : 262.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart